Bikinin 98%

Supplier: Ambeed
Warning

Synonyms: 4-[(5-Bromo-2-pyridyl)amino]-4-oxobutyric acid, 4-((5-Bromopyridin-2-yl)amino)-4-oxobutanoic acid

A176791-100MG A176791-10MG A176791-50MG A176791-1G A176791-5G
76872-592EA 31 USD
76872-592 76872-594 76872-596 77885-371 77885-372
Bikinin 98%
Bikinin

Precaución: for Research Use Only

Formula: C₉H₉BrN₂O₃
MDL Number: MFCD00116442
CAS Number: 188011-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR