Methyl 3-(2,2,2-trifluoroacetamido)thiophene-2-carboxylate 97%

Supplier: Ambeed
Warning

A175616-1G A175616-25G
76872-570EA 18.04 USD
76872-570 76872-572
Methyl 3-(2,2,2-trifluoroacetamido)thiophene-2-carboxylate 97%
Methyl 3-(2,2,2-trifluoroacetamido)thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₃S
MW: 253.2 g/mol
MDL Number: MFCD00103374
CAS Number: 79128-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR