2-Chloro-6-methoxypyridin-4-amine 95%
Supplier: Ambeed
|
Warning
|
A175552-100MG
A175552-250MG
A175552-1G
76872-566EA
30
USD
76872-566
76872-568
77593-110
2-Chloro-6-methoxypyridin-4-amine 95%
2-Chloro-6-methoxypyridin-4-amine
Precaución: for Research Use Only
Formula:
C₆H₇ClN₂O |
MDL Number:
MFCD13189324 CAS Number: 1008304-85-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...