9-Phenyl-3,3′-bicarbazole 97%

Supplier: Ambeed
Warning

Synonyms: 9-Phenyl-3,3'-bicarbazolyl

A175495-10G A175495-1G A175495-250MG
76872-560EA 79.57 USD
76872-560 76872-562 76872-564
9-Phenyl-3,3′-bicarbazole 97%
9-Phenyl-3,3′-bicarbazole

Precaución: for Research Use Only

Formula: C₃₀H₂₀N₂
MW: 408.493 g/mol
MDL Number: MFCD22581311
CAS Number: 1060735-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR