5-Bromo-1H-pyrrolo[2,3-b]pyridine-2,3-dione 95%

Supplier: Ambeed
Warning

A175403-100MG A175403-1G A175403-250MG
76872-556EA 45.62 USD
76872-556 76872-558 77593-108
5-Bromo-1H-pyrrolo[2,3-b]pyridine-2,3-dione 95%
5-Bromo-1H-pyrrolo[2,3-b]pyridine-2,3-dione

Precaución: for Research Use Only

Formula: C₇H₃BrN₂O₂
MW: 227.01 g/mol
MDL Number: MFCD13193450
CAS Number: 149142-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR