5-Amino-1-methyl-1H-pyrazole-4-carboxamide 96%

Supplier: Ambeed
Warning

A174858-1G A174858-250MG A174858-5G
76872-550EA 53.05 USD
76872-550 76872-552 77567-580
5-Amino-1-methyl-1H-pyrazole-4-carboxamide 96%
5-Amino-1-methyl-1H-pyrazole-4-carboxamide

Precaución: For research use only.

Formula: C₅H₈N₄O
MW: 140.15 g/mol
MDL Number: MFCD00111808
CAS Number: 18213-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR