4-(4-Methylpiperazin-1-yl)benzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Carboxyphenyl)-4-methylpiperazine, 4-(4-Methylpiperazinyl)benzoic acid

A174327-100G A174327-10G A174327-1G A174327-25G A174327-5G
76872-534EA 227 USD
76872-534 76872-536 76872-538 76872-540 76872-542
4-(4-Methylpiperazin-1-yl)benzoic acid 95%
4-(4-Methylpiperazin-1-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O₂
MDL Number: MFCD02682063
CAS Number: 86620-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR