2-Methyl-l-proline methyl ester hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (S)-Methyl 2-methylpyrrolidine-2-carboxylate hydrochloride

A174049-250MG A174049-5G
76872-526EA 13 USD
76872-526 77567-566
2-Methyl-l-proline methyl ester hydrochloride 97%
2-Methyl-l-proline methyl ester hydrochloride

Precaución: For research use only.

Formula: C₇H₁₃NO₂·ClH
MDL Number: MFCD22570171
CAS Number: 220060-08-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR