D-Tyrosinol hydrochloride 95%

Supplier: Ambeed
Warning

A173994-1G A173994-250MG A173994-5G
76872-520EA 114 USD
76872-520 76872-522 76872-524
D-Tyrosinol hydrochloride 95%
D-Tyrosinol hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₄ClNO₂
MW: 203.66 g/mol
MDL Number: MFCD01863503
CAS Number: 40829-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR