2-(Methoxycarbonyl)cyclopropane-1-carboxylic acid 98%

Supplier: Ambeed
Warning

A173927-5G A173927-1G A173927-250MG
76872-516EA 99 USD
76872-516 77611-692 77883-335
2-(Methoxycarbonyl)cyclopropane-1-carboxylic acid 98%
2-(Methoxycarbonyl)cyclopropane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₈O₄
MDL Number: MFCD16817526
CAS Number: 13279-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR