trans-4-(4-Morpholinyl)cyclohexanol 95%

Supplier: Ambeed
Warning

Synonyms: trans-4-Morpholinocyclohexanol

A173793-1G A173793-250MG
76872-512EA 113.52 USD
76872-512 76872-514
trans-4-(4-Morpholinyl)cyclohexanol 95%
trans-4-(4-Morpholinyl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₂
MW: 185.263 g/mol
MDL Number: MFCD20259655
CAS Number: 1228947-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR