2-(Difluoromethoxy)benzeneboronic acid, pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 2-(2-(Difluoromethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-[2-(Difluoromethoxy)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A173355-100MG A173355-1G A173355-250MG A173355-5G
76872-494EA 58.35 USD
76872-494 76872-496 76872-498 76872-500
2-(Difluoromethoxy)benzeneboronic acid, pinacol ester 95%
2-(Difluoromethoxy)benzeneboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BF₂O₃
MW: 270.08 g/mol
MDL Number: MFCD16996403
CAS Number: 960067-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR