(R)-(+)-2-Bromo-α-methylbenzyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: (R)-(+)-1-(2-Bromophenyl)ethanol, (R)-2-Bromo-alpha-methylbenzyl Alcohol

A172330-100MG A172330-1G A172330-250MG A172330-5G
76872-470EA 5 USD
76872-470 76872-472 76872-474 76872-476
(R)-(+)-2-Bromo-α-methylbenzyl alcohol 95%
(R)-(+)-2-Bromo-α-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MDL Number: MFCD00216591
CAS Number: 76116-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR