(Z)-2-(2-((tert-Butoxycarbonyl)amino)thiazol-4-yl)pent-2-enoic acid 97%

Supplier: Ambeed
Warning

A172081-100G A172081-10G A172081-1G A172081-25G A172081-5G
76872-448EA 144 USD
76872-448 76872-450 76872-452 76872-454 76872-456
(Z)-2-(2-((tert-Butoxycarbonyl)amino)thiazol-4-yl)pent-2-enoic acid 97%
(Z)-2-(2-((tert-Butoxycarbonyl)amino)thiazol-4-yl)pent-2-enoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₄S
MDL Number: MFCD08458237
CAS Number: 86978-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR