5-Bromo-1,3-dichloro-2-methylbenzene 98%

Supplier: Ambeed
Warning

A171096-10G A171096-1G A171096-250MG A171096-5G
76872-440EA 285.39 USD
76872-440 76872-442 76872-444 76872-446
5-Bromo-1,3-dichloro-2-methylbenzene 98%
5-Bromo-1,3-dichloro-2-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂
MW: 239.93 g/mol
MDL Number: MFCD11227146
CAS Number: 204930-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR