2-Bromo-5-nitrophenol 97%

Supplier: Ambeed
Warning

A170954-100G A170954-10G A170954-1G A170954-250MG A170954-5G A170954-25G
76872-424EA 507.12 USD
76872-424 76872-426 76872-428 76872-430 76872-432 77593-066
2-Bromo-5-nitrophenol 97%
2-Bromo-5-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₄BrNO₃
MW: 218.0100 g/mol
MDL Number: MFCD00466299
CAS Number: 52427-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR