4-[N-Methyl-N-(4-methoxybenzyl)sulfamoyl]benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(N-(4-Methoxybenzyl)-N-methylsulphamoyl)phenyl)boronic acid

A170603-1G A170603-5G
76872-410EA 41.38 USD
76872-410 76872-412
4-[N-Methyl-N-(4-methoxybenzyl)sulfamoyl]benzeneboronic acid 98%
4-[N-Methyl-N-(4-methoxybenzyl)sulfamoyl]benzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₈BNO₅S
MW: 335.183 g/mol
MDL Number: MFCD08689488
CAS Number: 913835-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR