(S)-2-(Pyrrolidin-2-yl)propan-2-ol 97%

Supplier: Ambeed
Warning

A170491-100MG A170491-250MG A170491-1G
76872-406EA 43.5 USD
76872-406 76872-408 77611-662
(S)-2-(Pyrrolidin-2-yl)propan-2-ol 97%
(S)-2-(Pyrrolidin-2-yl)propan-2-ol

Precaución: for Research Use Only

Formula: C₇H₁₅NO
MW: 129.20 g/mol
MDL Number: MFCD11975452
CAS Number: 92053-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR