Methyl 3-aminocyclobutanecarboxylate hydrochloride 98%

Supplier: Ambeed
Warning

A170386-1G A170386-5G A170386-100MG A170386-25G
76872-404EA 85.94 USD
76872-404 77611-660 77671-416 77671-418
Methyl 3-aminocyclobutanecarboxylate hydrochloride 98%
Methyl 3-aminocyclobutanecarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO₂
MDL Number: MFCD17015886
CAS Number: 1354940-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR