2-Amino-4-chloro-5-methylbenzenesulfonic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-2-chlorotoluene-5-sulphonic acid

A170117-100G A170117-10G A170117-25G A170117-500G A170117-5G
76872-388EA 37.14 USD
76872-388 76872-390 76872-392 76872-394 76872-396
2-Amino-4-chloro-5-methylbenzenesulfonic acid 97%
2-Amino-4-chloro-5-methylbenzenesulphonic acid

Precaución: for Research Use Only

Formula: C₇H₈ClNO₃S
MW: 221.6613 g/mol
MDL Number: MFCD00035771
CAS Number: 88-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR