4,4,5,5-Tetramethyl-2-(2-methyl-1-propenyl)-1,3,2-dioxaborolane 97%

Supplier: Ambeed
Warning

Synonyms: (2-Methyl-1-propenyl)boronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-(2-methylprop-1-en-1-yl)-1,3,2-dioxaborolane

A170089-100G A170089-10G A170089-1G A170089-25G A170089-500G A170089-5G
76872-376EA 401.03 USD
76872-376 76872-378 76872-380 76872-382 76872-384 76872-386
4,4,5,5-Tetramethyl-2-(2-methyl-1-propenyl)-1,3,2-dioxaborolane 97%
4,4,5,5-Tetramethyl-2-(2-methyl-1-propenyl)-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₀H₁₉BO₂
MW: 182.07 g/mol
MDL Number: MFCD09842762
CAS Number: 126689-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR