2-Chloro-3-hydroxy-4-picoline 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-methylpyridin-3-ol

A169978-1G A169978-250MG
76872-368EA 153.84 USD
76872-368 76872-370
2-Chloro-3-hydroxy-4-picoline 97%
2-Chloro-3-hydroxy-4-picoline

Precaución: for Research Use Only

Formula: C₆H₆ClNO
MW: 143.5700 g/mol
MDL Number: MFCD04112564
CAS Number: 884494-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR