Ethyl-2-amino-2-cyanoacetate mono(4-methylbenzenesulfonate) 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-amino-2-cyanoacetate 4-methylbenzenesulphonate

A169895-100MG A169895-10G A169895-1G A169895-250MG A169895-25G A169895-5G
76872-356EA 10.61 USD
76872-356 76872-358 76872-360 76872-362 76872-364 76872-366
Ethyl-2-amino-2-cyanoacetate mono(4-methylbenzenesulfonate) 97%
Ethyl 2-amino-2-cyanoacetate mono(4-methylbenzenesulphonate)

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O₅S
MW: 300.3308 g/mol
MDL Number: MFCD00666813
CAS Number: 37842-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR