2,6-Dimethylterephthalic acid 96%

Supplier: Ambeed
Warning

A169609-100MG A169609-1G A169609-250MG
76872-346EA 33.95 USD
76872-346 76872-348 76872-350
2,6-Dimethylterephthalic acid 96%
2,6-Dimethylterephthalic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.18 g/mol
MDL Number: MFCD00182548
CAS Number: 80238-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR