N-[2-(Trimethylsilyl)ethoxycarbonyloxy]succinimide 97%
Supplier: Ambeed
|
Warning
|
A169521-100G
A169521-10G
A169521-1G
A169521-250MG
A169521-25G
A169521-500G
A169521-5G
76872-332EA
437
USD
76872-332
76872-334
76872-336
76872-338
76872-340
76872-342
76872-344
N-[2-(Trimethylsilyl)ethoxycarbonyloxy]succinimide 97%
N-[2-(Trimethylsilyl)ethoxycarbonyloxy]succinimide
Precaución: for Research Use Only
Formula:
C₁₀H₁₇NO₅Si |
MDL Number:
MFCD02683467 CAS Number: 78269-85-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...