3-Fluoro-2-methylpyridin-4-amine 95%

Supplier: Ambeed
Warning

A169249-1G A169249-250MG A169249-50MG
76872-328EA 320 USD
76872-328 76872-330 77885-332
3-Fluoro-2-methylpyridin-4-amine 95%
3-Fluoro-2-methylpyridin-4-amine

Precaución: for Research Use Only

Formula: C₆H₇FN₂
MDL Number: MFCD18261569
CAS Number: 15931-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR