Fmoc-Lys(Tfa)-OH 98%

Supplier: Ambeed
Warning

A168618-100G A168618-1G A168618-250MG A168618-25G A168618-5G
76872-310EA 174 USD
76872-310 76872-312 76872-314 76872-316 76872-318
Fmoc-Lys(Tfa)-OH 98%
Fmoc-Lys(Tfa)-OH

Precaución: for Research Use Only

Formula: C₂₃H₂₃F₃N₂O₅
MDL Number: MFCD00153360
CAS Number: 76265-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR