Methyl 4-chloro-2-methoxynicotinate 98%

Supplier: Ambeed
Warning

A168480-250MG A168480-5G
76872-302EA 11 USD
76872-302 77883-303
Methyl 4-chloro-2-methoxynicotinate 98%
Methyl 4-chloro-2-methoxynicotinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₃
MDL Number: MFCD18257740
CAS Number: 1256826-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR