5-Bromo-4-fluorobenzo[d][1,3]dioxole 95%

Supplier: Ambeed
Warning

A168420-100MG A168420-1G A168420-250MG A168420-5G
76872-294EA 72.15 USD
76872-294 76872-296 76872-298 76872-300
5-Bromo-4-fluorobenzo[d][1,3]dioxole 95%
5-Bromo-4-fluorobenzo[d][1,3]dioxole

Precaución: for Research Use Only

Formula: C₇H₄BrFO₂
MDL Number: MFCD16295053
CAS Number: 1226808-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR