6-Bromo-5-fluoro-1H-benzo[d][1,2,3]triazole 95%

Supplier: Ambeed
Warning

A168166-1G A168166-250MG
76872-276EA 60 USD
76872-276 76872-278
6-Bromo-5-fluoro-1H-benzo[d][1,2,3]triazole 95%
6-Bromo-5-fluoro-1H-benzo[d][1,2,3]triazole

Precaución: for Research Use Only

Formula: C₆H₃BrFN₃
MDL Number: MFCD16660325
CAS Number: 1242336-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR