4-(4-Bromophenyl)-1-(methylsulfonyl)piperidine 97%

Supplier: Ambeed
Warning

A167921-1G A167921-250MG
76872-272EA 10 USD
76872-272 76872-274
4-(4-Bromophenyl)-1-(methylsulfonyl)piperidine 97%
4-(4-Bromophenyl)-1-(methylsulfonyl)piperidine

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrNO₂S
MDL Number: MFCD23701556
CAS Number: 622386-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR