1-Aminopyrrolidin-2-one hydrochloride 95%

Supplier: Ambeed
Warning

A167782-100MG A167782-10G A167782-1G A167782-5G A167782-250MG
76872-264EA 33.95 USD
76872-264 76872-266 76872-268 76872-270 77671-374
1-Aminopyrrolidin-2-one hydrochloride 95%
1-Aminopyrrolidin-2-one hydrochloride

Precaución: for Research Use Only

Formula: C₄H₈N₂O·ClH
MW: 136.58 g/mol
MDL Number: MFCD09258892
CAS Number: 20386-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR