5-Nitro-3-(trifluoromethyl)pyridin-2-amine 96%

Supplier: Ambeed
Warning

A167408-100MG A167408-250MG
76872-248EA 41 USD
76872-248 76872-250
5-Nitro-3-(trifluoromethyl)pyridin-2-amine 96%
5-Nitro-3-(trifluoromethyl)pyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₄F₃N₃O₂
MW: 207.11 g/mol
MDL Number: MFCD18382741
CAS Number: 1121056-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR