4-Bromo-5-chloro-2-methoxypyridine 98%

Supplier: Ambeed
Warning

A167184-100MG A167184-1G A167184-250MG
76872-242EA 87 USD
76872-242 76872-244 76872-246
4-Bromo-5-chloro-2-methoxypyridine 98%
4-Bromo-5-chloro-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClNO
MW: 222.47 g/mol
MDL Number: MFCD16609882
CAS Number: 1211534-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR