4-(Difluoromethyl)benzeneboronic acid 97%

Supplier: Ambeed
Danger

Synonyms: (4-(Difluoromethyl)phenyl)boronic acid, 4-Boronobenzal fluoride

A166770-100MG A166770-1G A166770-250MG A166770-5G
76872-228EA 7 USD
76872-228 76872-230 76872-232 76872-234
4-(Difluoromethyl)benzeneboronic acid 97%
4-(Difluoromethyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₂
MDL Number: MFCD09056705
CAS Number: 946525-43-5
UN: 2921
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR