Isopropyl-4-hydroxypiperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Isopropyl 4-hydroxypiperidine-1-carboxylate

A166739-1G A166739-25G A166739-5G
76872-222EA 10.61 USD
76872-222 76872-224 76872-226
Isopropyl-4-hydroxypiperidine-1-carboxylate 98%
Isopropyl-4-hydroxypiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₇NO₃
MW: 187.2400 g/mol
MDL Number: MFCD08460168
CAS Number: 832715-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR