4,4,5,5-Tetramethyl-2-(2-(naphthalen-2-yl)phenyl)-1,3,2-dioxaborolane 95%
Supplier: Ambeed
|
Warning
|
A166705-250MG
A166705-5G
A166705-100MG
A166705-1G
76872-220EA
36
USD
76872-220
77671-354
77883-299
77883-300
4,4,5,5-Tetramethyl-2-(2-(naphthalen-2-yl)phenyl)-1,3,2-dioxaborolane 95%
4,4,5,5-Tetramethyl-2-(2-(naphthalen-2-yl)phenyl)-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₂₂H₂₃BO₂ |
MDL Number:
MFCD22421637 CAS Number: 1062555-59-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...