5-Cyano-1H-indazole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A166463-1G A166463-250MG A166463-5G
76872-214EA 191 USD
76872-214 76872-216 77883-298
5-Cyano-1H-indazole-3-carboxylic acid 97%
5-Cyano-1H-indazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅N₃O₂
MDL Number: MFCD07378942
CAS Number: 885520-03-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR