4-Bromo-5-fluoro-2-methylbenzonitrile 97%

Supplier: Ambeed
Warning

A166366-1G A166366-250MG A166366-5G
76872-208EA 22 USD
76872-208 76872-210 76872-212
4-Bromo-5-fluoro-2-methylbenzonitrile 97%
4-Bromo-5-fluoro-2-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrFN
MDL Number: MFCD07782074
CAS Number: 916792-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR