4-(4-Fluorobenzyl)piperidine 97%

Supplier: Ambeed
Warning

A166353-1G A166353-5G
76872-206EA 131 USD
76872-206 77885-317
4-(4-Fluorobenzyl)piperidine 97%
4-(4-Fluorobenzyl)piperidine

Precaución: for Research Use Only

Formula: C₁₂H₁₆FN
MDL Number: MFCD03839825
CAS Number: 92822-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR