2-(3-Bromo-5-isobutoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-isobutoxyphenylboronic acid pinacol ester

A166346-100MG A166346-1G A166346-250MG A166346-5G
76872-198EA 37.14 USD
76872-198 76872-200 76872-202 76872-204
2-(3-Bromo-5-isobutoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(3-Bromo-5-isobutoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₆H₂₄BBrO₃
MW: 355.08 g/mol
MDL Number: MFCD12546587
CAS Number: 1218789-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR