(2-Chloropyridin-4-yl)methanamine hydrochloride 95%

Supplier: Ambeed
Warning

A166155-1G A166155-250MG
76872-184EA 68.96 USD
76872-184 76872-186
(2-Chloropyridin-4-yl)methanamine hydrochloride 95%
(2-Chloropyridin-4-yl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₈Cl₂N₂
MW: 179.05 g/mol
MDL Number: MFCD21362508
CAS Number: 916210-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR