Ethyl 10-Bromo-9-anthracenecarboxylate 97%

Supplier: Ambeed
Warning

A164059-100MG A164059-1G A164059-250MG A164059-5G
76872-120EA 17 USD
76872-120 76872-122 76872-124 76872-126
Ethyl 10-Bromo-9-anthracenecarboxylate 97%
Ethyl 10-Bromo-9-anthracenecarboxylate

Precaución: for Research Use Only

Formula: C₁₇H₁₃BrO₂
MDL Number: MFCD22571679
CAS Number: 1089318-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR