2-(Ethoxycarbonyl)-1,3-thiazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Ethoxycarbonyl)thiazole-4-carboxylic acid

A162858-100MG A162858-1G A162858-250MG A162858-5G A162858-25G
76872-092EA 16.98 USD
76872-092 76872-094 76872-096 77592-980 77671-298
2-(Ethoxycarbonyl)-1,3-thiazole-4-carboxylic acid 97%
2-(Ethoxycarbonyl)-1,3-thiazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₄S
MW: 201.2 g/mol
MDL Number: MFCD11110995
CAS Number: 911466-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR