[(Trimethylsilyl)ethynyl]boronic acid, pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-[(trimethylsilyl)ethynyl]-1,3,2-dioxaborolane, Trimethyl((4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethynyl)silane, [(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)ethynyl]trimethylsilane

A162316-100MG A162316-1G A162316-250MG A162316-5G
76872-078EA 20 USD
76872-078 76872-080 76872-082 76872-084
[(Trimethylsilyl)ethynyl]boronic acid, pinacol ester 95%
[(Trimethylsilyl)ethynyl]boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₂₁BO₂Si
MDL Number: MFCD13182130
CAS Number: 159087-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR