4-(4-Fluorophenylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-((4-Fluorophenyl)carbamoyl)phenyl)boronic acid, 4-[(4-Fluorophenyl)aminocarbonyl]benzeneboronic acid

A162311-25G A162311-5G
76872-074EA 54 USD
76872-074 76872-076
4-(4-Fluorophenylcarbamoyl)phenylboronic acid 98%
4-(4-Fluorophenylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BFNO₃
MDL Number: MFCD06659878
CAS Number: 850568-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR