tert-Butyl 2,2-dimethyl-4-oxopiperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A162110-100MG A162110-10G A162110-1G A162110-250MG A162110-25G A162110-5G
76872-050EA 35.01 USD
76872-050 76872-052 76872-054 76872-056 76872-058 76872-060
tert-Butyl 2,2-dimethyl-4-oxopiperidine-1-carboxylate 97%
tert-Butyl 2,2-dimethyl-4-oxopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₃
MW: 227.30 g/mol
MDL Number: MFCD18914358
CAS Number: 346593-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR