3-(Aminomethyl)-4,6-dimethyl-1,2-dihydropyridin-2-one 97%

Supplier: Ambeed
Warning

A161991-1G A161991-250MG A161991-25G A161991-5G
76872-042EA 16 USD
76872-042 76872-044 76872-046 76872-048
3-(Aminomethyl)-4,6-dimethyl-1,2-dihydropyridin-2-one 97%
3-(Aminomethyl)-4,6-dimethyl-1,2-dihydropyridin-2-one

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O
MDL Number: MFCD06213260
CAS Number: 771579-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR