N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine 97%

Supplier: Ambeed
Warning

Synonyms: N-[(S)-(+)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine, NEPA, N-[(S)-1-Carboxyethyl]-L-homophenylalanine Ethyl Ester, (S)-2-(((S)-1-Ethoxy-1-oxo-4-phenylbutan-2-yl)amino)propanoic acid, N-(1S)-Ethoxycarbonyl-3-phenylpropyl-(S)-alanine

A161643-100G A161643-25G A161643-5G A161643-10G A161643-500G
76872-038EA 55 USD
76872-038 76872-040 77671-288 77883-270 77883-271
N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine 97%
N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₄
MW: 279.34 g/mol
MDL Number: MFCD00209976
CAS Number: 82717-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR