2-Bromo-5-nitro-3-(trifluoromethyl)pyridine 97%

Supplier: Ambeed
Warning

A161632-10G A161632-1G A161632-250MG A161632-5G
76872-030EA 185 USD
76872-030 76872-032 76872-034 76872-036
2-Bromo-5-nitro-3-(trifluoromethyl)pyridine 97%
2-Bromo-5-nitro-3-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂BrF₃N₂O₂
MW: 270.9915 g/mol
MDL Number: MFCD17169962
CAS Number: 956104-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR